Få mere at vide
N,N-Dimethylaniline-d3 (N-methyl-d3), CDN
Artikelnummer. 30314366
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30314366 0.25 g -
30311865 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314366 Leverandør CDN Leverandørnr. CDND62160.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Dimethylaniline-D3 (N-methyl-D3)
CAS 88889-00-3
Molekylær formel C8 D3 H8 N
Mængde 0.25 g
Synonym Benzenamine, N-methyl-N-(methyl-d3)- (9 CI, ACI), N-Methyl-N-(methyl-d3)benzenamine (ACI), N-Methyl-N-(methyl-d3)aniline, N,N-Dimethylaniline-d3 (N-methyl-d3), N,N-Dimethylaniline-D3 (N-methyl-D3)
InChI formel InChI=1 S/C8H11N/c1-9(2)8-6-4-3-5-7-8/h3-7 H,1-2H3/i1D3
SMIL [2 H]C([2 H])([2 H])N(C)c1ccccc1
IUPAC navn N-methyl-N-(trideuteriomethyl)aniline
Molekylvægt (g/mol) 124.198
Formel vægt 124.108 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity