Få mere at vide
N,N-Dimethylaniline-d11, TRC
Artikelnummer. 30465187
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30465187 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30465187 Leverandør Toronto Research Chemicals Leverandørnr. TRCN46410250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Dimethylaniline-d11
CAS 85785-00-8
Molekylær formel C8 D11 N
Mængde 50 mg
Synonym Benzen-d5-amine','N,N-di(methyl-d3)- (9CI), Aniline-d5','N,N-di(methyl-d3)- (6CI), N,N-Dimethylaniline-d11, N,N-Dimethylaniline-D11, N,N-Di(methyl-d3)aniline-d5
InChI formel InChI=1S/C8H11N/c1-9(2)8-6-4-3-5-7-8/h3-7H,1-2H3/i1D3,2D3,3D,4D,5D,6D,7D
SMIL [2H]c1c([2H])c([2H])c(N(C([2H])([2H])[2H])C([2H])([2H])[2H])c([2H])c1[2H]
IUPAC navn 2,3,4,5,6-pentadeuterio-N,N-bis(trideuteriomethyl)aniline
Molekylvægt (g/mol) 132.25
Formel vægt 132.1582