Få mere at vide
N,N-Dimethylaniline-d11, CDN
Artikelnummer. 30321975
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321975 0.25 g -
30310809 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321975 Leverandør CDN Leverandørnr. CDND57300.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Dimethylaniline-d11
CAS 85785-00-8
Molekylær formel C8 D11 N
Mængde 0.25 g
InChI formel InChI=1 S/C8H11N/c1-9(2)8-6-4-3-5-7-8/h3-7 H,1-2H3/i1D3,2D3,3 D,4 D,5 D,6 D,7 D
SMIL [2 H]c1c([2 H])c([2 H])c(N(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])c([2 H])c1[2 H]
IUPAC navn 2,3,4,5,6-pentadeuterio-N,N-bis(trideuteriomethyl)aniline
Molekylvægt (g/mol) 132.25
Formel vægt 132.1582 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity