Få mere at vide
N,N-Dimethylamidino Urea, TRC
Artikelnummer. 30374886
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30374886 250 mg
30369586 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30374886 Leverandør Toronto Research Chemicals Leverandørnr. TRCD471205250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 98026-16-5
Molekylær formel C4H10N4O
Molekylvægt (g/mol) 130.15
Synonym N-[(Dimethylamino)iminomethyl]Urea, N3-Desamidino,N3-carbamoyl Metformin, N3-Desamidino,N3-hydroxyimidoyl Metformin, Metformin Hydroxy Analog
IUPAC navn (N,N-dimethylcarbamimidoyl)urea
SMIL NC(NC(N(C)C)=N)=O≫N=C(O)NC(N(C)C)=N
Tekniske data
Kemisk navn eller materiale N,N-Dimethylamidino Urea
Mængde 250 mg
InChI formel InChI=1S/2C4H10N4O/c2*1-8(2)3(5)7-4(6)9/h2*1-2H3,(H4,5,6,7,9)
Formel vægt 130.09