missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number 11330024
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
3575-32-4
C8H14Cl2N2
209.114
MFCD00012971
BZJPIQKDEGXVFG-UHFFFAOYSA-N
n1,n1-dimethylbenzene-1,3-diamine dihydrochloride, n,n-dimethyl-m-phenylenediamine dihydrochloride, m-dimethylamino aniline, n,n-dimethyl-1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine, n,n-dimethyl-, dihydrochloride, n,n-dimethylbenzene-1,3-diamine dihydrochloride, 3-amino-n,n-dimethylaniline, 1,3-benzenediamine, n1,n1-dimethyl-, hydrochloride 1:2, pubchem14991, acmc-20a5k9
77124
3-N,3-N-dimethylbenzene-1,3-diamine;dihydrochloride
CN(C)C1=CC=CC(=C1)N.Cl.Cl
This item is not returnable. View return policy
3575-32-4
C8H14Cl2N2
209.114
MFCD00012971
BZJPIQKDEGXVFG-UHFFFAOYSA-N
n1,n1-dimethylbenzene-1,3-diamine dihydrochloride, n,n-dimethyl-m-phenylenediamine dihydrochloride, m-dimethylamino aniline, n,n-dimethyl-1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine, n,n-dimethyl-, dihydrochloride, n,n-dimethylbenzene-1,3-diamine dihydrochloride, 3-amino-n,n-dimethylaniline, 1,3-benzenediamine, n1,n1-dimethyl-, hydrochloride 1:2, pubchem14991, acmc-20a5k9
77124
3-N,3-N-dimethylbenzene-1,3-diamine;dihydrochloride
CN(C)C1=CC=CC(=C1)N.Cl.Cl

it can be used to produce N-(4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-yl)-formamide with 4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-ylamine. This reaction will need solvent tetrahydrofuran. it is applied as a selective formylating agent for one of two amino groups in such basic amino acids as ornithine or lysine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
it can be used to produce N-(4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-yl)-formamide with 4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-ylamine. This reaction will need solvent tetrahydrofuran. it is applied as a selective formylating agent for one of two amino groups in such basic amino acids as ornithine or lysine.

Solubility
Soluble in water, chloroform and dichloromethane. Slightly soluble in ethyl acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point 216°C (decomposition)
Quantity 1 g
UN Number UN2811
Beilstein 3694384
Solubility Information Soluble in water,chloroform and dichloromethane. Slightly soluble in ethyl acetate.
Formula Weight 209.12
Percent Purity 99%
Chemical Name or Material N,N-Dimethyl-m-phenylenediamine dihydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'