Få mere at vide
(N,N)-Dimethyl)ethyl Laurate, TRC
Artikelnummer. 30377102
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30377102 50 mg
30375402 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30377102 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47295050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 34839-10-6
Molekylær formel C16H33NO2
Molekylvægt (g/mol) 271.44
Synonym Dodecanoic Acid 2-(Dimethylamino)ethyl Ester, 2-(N,N-Dimethylamino)ethyl Dodecanoate, N-[2-(Dodecanoyloxy)ethyl]-N,N-dimethylamine, NSC 405542
SMIL CCCCCCCCCCCC(=O)OCCN(C)C
Tekniske data
Kemisk navn eller materiale (N,N)-(Dimethyl)ethyl Laurate
Mængde 50 mg
InChI formel InChI=1S/C16H33NO2/c1-4-5-6-7-8-9-10-11-12-13-16(18)19-15-14-17(2)3/h4-15H2,1-3H3
Formel vægt 271.2511