Få mere at vide
N,N-Dimethyl-d6-formamide, CDN
Artikelnummer. 30320825
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320825 0.5 g 0.5g
30320713 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320825 Leverandør CDN Leverandørnr. CDND08900.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Dimethylformamide-D6 (N,N-dimethyl-D6)
CAS 185990-36-7
Molekylær formel C3 D6 H N O
Mængde 0.5 g
Synonym Formamide, N,N-di(methyl-d3)- (9 CI), DMF-d6, N,N-Di(methyl-d3)formamide, N,N-Dimethyl-d6-formamide
InChI formel InChI=1 S/C3H7NO/c1-4(2)3-5/h3H,1-2H3/i1D3,2D3
SMIL [2 H]C([2 H])([2 H])N(C=O)C([2 H])([2 H])[2 H]
IUPAC navn N,N-bis(trideuteriomethyl)formamide
Molekylvægt (g/mol) 79.1308
Formel vægt 79.0904 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity