Få mere at vide
N,N-Dimethyl-d6-4-nitrosoaniline, CDN
Artikelnummer. 30313156
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30313156 0.1 g 0.1g
30309822 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313156 Leverandør CDN Leverandørnr. CDND74780.1G
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Dimethyl-4-nitrosoaniline-D6 (N,N-dimethyl-D6)
CAS 1398065-85-4
Molekylær formel C8 D6 H4 N2 O
Mængde 0.1 g
Synonym N,N-Dimethyl-4-nitrosoaniline-D6 (N,N-dimethyl-D6), N,N-Di(methyl-d3)-4-nitrosobenzenamine, 1-[Di(methyl-d3)amino]-4-nitrosobenzene, 4-(Dimethylamino-d6)nitrosobenzene, 4-Nitroso-N,N-di(methyl-d3)aminobenzene, 4-Nitroso-N,N-dimethylaniline-d6, N,N-Dimethyl-p-nitrosoaniline-d6, NDMA, p-(Dimethylamino)nitrosobenzene-d6, p-[N,N-Bis(trideuteriomethyl)amino]nitrosobenzene, p-NDMA-d6, p-Nitroso-N,N-dimethylaniline-d6
InChI formel InChI=1 S/C8H10N2O/c1-10(2)8-5-3-7(9-11)4-6-8/h3-6 H,1-2H3/i1D3,2D3
SMIL [2 H]C([2 H])([2 H])N(c1ccc(cc1)N=O)C([2 H])([2 H])[2 H]
IUPAC navn 4-nitroso-N,N-bis(trideuteriomethyl)aniline
Molekylvægt (g/mol) 156.215
Formel vægt 156.117 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity