Få mere at vide
N,N-Dimethyl-d3-glycine HCl (N-methyl-d3), CDN
Artikelnummer. 30309590
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30309590 0.25 g -
30321875 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309590 Leverandør CDN Leverandørnr. CDND70240.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Dimethylglycine-D3 (N-methyl-D3) Hydrochloride
CAS 1219795-14-8
Molekylær formel C4 D3 H6 N O2 . Cl H
Mængde 0.25 g
Synonym (Dimethylamino)acetic acid-d3 hydrochloride, 2-[N-Methyl-N-(methyl-d3)amino]acetic acid hydrochloride, Dimethylglycine-d3 hydrochloride, N-Methyl-N-(methyl-d3)aminoacetic acid hydrochloric acid, N,N-Dimethylglycine-d3 hydrochloride, N,N-Dimethylglycine-D3 (N-methyl-D3) hydrochloride
InChI formel InChI=1 S/C4H9NO2.ClH/c1-5(2)3-4(6)7;/h3H2,1-2H3,(H,6,7);1 H/i1D3;
SMIL Cl.[2 H]C([2 H])([2 H])N(C)CC(=O)O
IUPAC navn 2-[methyl(trideuteriomethyl)amino]acetic acid;hydrochloride
Molekylvægt (g/mol) 142.599
Formel vægt 142.059 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity