Få mere at vide
N,N-Dimethyl-5-(phenylmethoxy)-1H-indole-3-ethan-a,a,b,b-d4-amine, TRC
Artikelnummer. 30369232
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30369232 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30369232 Leverandør Toronto Research Chemicals Leverandørnr. TRCD688300100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Dimethyl-5-(phenylmethoxy)-1H-indole-3-ethan-alpha,alpha,beta,beta-d4-amine
CAS 688301-42-0
Molekylær formel C19H18D4N2O
Mængde 100 mg
Synonym Uracil Lyxonucleoside-13C,15N2 5'-Phosphate
InChI formel InChI=1S/C19H22N2O/c1-21(2)11-10-16-13-20-19-9-8-17(12-18(16)19)22-14-15-6-4-3-5-7-15/h3-9,12-13,20H,10-11,14H2,1-2H3/i10D2,11D2
SMIL [2H]C([2H])(N(C)C)C([2H])([2H])c1c[nH]c2ccc(OCc3ccccc3)cc12
IUPAC navn 1,1,2,2-tetradeuterio-N,N-dimethyl-2-(5-phenylmethoxy-1H-indol-3-yl)ethanamine
Molekylvægt (g/mol) 298.42
Formel vægt 298.1983