Få mere at vide
N,N-Dimethyl-4-hydroxymethoxy Clomiphene (~90%, contains ~10% Z-isomer), TRC
Artikelnummer. 30006956
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006956 5 mg
30006097 10 mg
30005378 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006956 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9729385MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 204123-74-0
Molekylær formel C25H26ClNO3
Molekylvægt (g/mol) 423.93
Synonym (E)-4-[2-Chloro-1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-(4-methoxyphenyl)ethenyl]phenol
IUPAC navn 4-[(E)-2-chloro-1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-(4-methoxyphenyl)vinyl]phenol
SMIL COc1ccc(cc1)\C(=C(\c2ccc(O)cc2)/c3ccc(OCCN(C)C)cc3)\Cl
Tekniske data
Kemisk navn eller materiale 4-Hydroxymethoxy Clomiphene
Mængde 5 mg
InChI formel InChI=1S/C25H26ClNO3/c1-27(2)16-17-30-23-14-6-19(7-15-23)24(18-4-10-21(28)11-5-18)25(26)20-8-12-22(29-3)13-9-20/h4-15,28H,16-17H2,1-3H3/b25-24+
Formel vægt 423.1601