Få mere at vide
N,N'-Dimethyl-3,3'-dithiodipropionamide-d6, TRC
Artikelnummer. 30375239
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30375239 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30375239 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47270710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N'-Dimethyl-3,3'-dithiodipropionamide-d6
CAS 2374866-79-0
Molekylær formel C8H10D6N2O2S2
Mængde 10 mg
Synonym 3,3'-Dithiobis[N-(methyl-d3)propanamide]
InChI formel InChI=1S/C8H16N2O2S2/c1-9-7(11)3-5-13-14-6-4-8(12)10-2/h3-6H2,1-2H3,(H,9,11)(H,10,12)/i1D3,2D3
SMIL O=C(CCSSCCC(NC([2H])([2H])[2H])=O)NC([2H])([2H])[2H]
IUPAC navn 3,3'-disulfanediylbis(N-(methyl-d3)propanamide)
Molekylvægt (g/mol) 242.39
Formel vægt 242.1