Få mere at vide
N,N-Dimethyl-2-methylene-Beta-alanine, TRC
Artikelnummer. 30372778
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30372778 1 g
30375530 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30372778 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4747651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5415-98-5
Molekylær formel C6H11NO2
Molekylvægt (g/mol) 129.16
Synonym 2-[(Dimethylamino)methyl]-2-propenoic Acid, 2-[(Dimethylamino)methyl]acrylic Acid, NSC 11325
SMIL CN(C)CC(=C)C(=O)O
Tekniske data
Kemisk navn eller materiale N,N-Dimethyl-2-methylene-beta-alanine
Mængde 1 g
InChI formel InChI=1S/C6H11NO2/c1-5(6(8)9)4-7(2)3/h1,4H2,2-3H3,(H,8,9)
Formel vægt 129.079