missing translation for 'onlineSavingsMsg'
Få mere at vide

N,N-Dimethyl-2-aminosulfonyl-4-formamidobenzamide, TRC

Artikelnummer. 30368872
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30368872 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30368872 Leverandør Toronto Research Chemicals Leverandørnr. TRCD89687025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-(Aminosulfonyl)-4-(formylamino)-N,N-dimethylbenzamide
CAS 173159-94-9
Molekylær formel C10 H13 N3 O4 S
Mængde 25 mg
Synonym Benzamide, 2-(aminosulfonyl)-4-(formylamino)-N,N-dimethyl-, 2-(Aminosulfonyl)-4-(formylamino)-N,N-dimethylbenzamide, 2-Dimethylaminocarbonyl-5-(formylamino)benzenesulfonamide, N,N-Dimethyl-2-aminosulfonyl-4-formamidobenzamide, AE F153745
InChI formel InChI=1S/C10H13N3O4S/c1-13(2)10(15)8-4-3-7(12-6-14)5-9(8)18(11,16)17/h3-6H,1-2H3,(H,12,14)(H2,11,16,17)
SMIL CN(C)C(=O)c1ccc(NC=O)cc1S(=O)(=O)N
IUPAC navn 4-formamido-N,N-dimethyl-2-sulfamoylbenzamide
Molekylvægt (g/mol) 271.2929
Formel vægt 271.0627
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.