Få mere at vide
N,N-Dimethyl-2-[7-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetamide, Mikromol™
Artikelnummer. 17902596
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17902596 100 mg
17991816 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17902596 Leverandør Mikromol Leverandørnr. MM0333.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N,N-Dimethyl-2-[7-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Dimethyl-2-[7-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetamide
CAS 1346600-85-8
Analyt- eller komponentnavne N,N-Dimethyl-2-[7-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetamide
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C19 H21 N3 O
Certificeringer/compliance ISO 17025
Synonym N,N-Dimethyl-2-[7-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetamide, Zolpidem Tartrate Imp. A (EP), 5-Methyl Zolpidem
InChI formel InChI=1S/C19H21N3O/c1-13-5-7-15(8-6-13)19-16(12-18(23)21(3)4)22-10-9-14(2)11-17(22)20-19/h5-11H,12H2,1-4H3
SMIL CN(C)C(=O)Cc1c(nc2cc(C)ccn12)c3ccc(C)cc3
IUPAC navn N,N-dimethyl-2-[7-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetamide
Molekylvægt (g/mol) 307.39
Mængde 100 mg
Formel vægt 307.1685 g/mol