Få mere at vide
N,N-Diisopropylamine-d14 Hydrochloride, TRC
Artikelnummer. 30371875
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30371875 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371875 Leverandør Toronto Research Chemicals Leverandørnr. TRCD455237100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Diisopropylamine-d14, Hydrochloride
CAS 1219803-88-9
Molekylær formel C6 2H14 H N . Cl H
Mængde 100 mg
Synonym N-[1-(Methyl-d3)ethyl-1,2,2,2-d4]-2-propan-1,1,1,2,3,3,3-d7-amine hydrochloride, Di-iso-propyl-d14-amine hydrochloride, Diisopropylamine-d14 hydrochloride, Diisopropylamine-D14 hydrochloride, N,N-Diisopropylamine-d14 hydrochloride
InChI formel InChI=1S/C6H15N.ClH/c1-5(2)7-6(3)4;/h5-7H,1-4H3;1H/i1D3,2D3,3D3,4D3,5D,6D;
SMIL Cl.[2H]C([2H])([2H])C([2H])(NC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1,1,1,2,3,3,3-heptadeuterio-N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)propan-2-amine;hydrochloride
Molekylvægt (g/mol) 151.74
Formel vægt 151.185