Få mere at vide
N,N-Diethylethanol-1,1,2,2-d4-amine, CDN
Artikelnummer. 30317367
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30317367 0.1 g 0.1g
30325677 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317367 Leverandør CDN Leverandørnr. CDND77160.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(Diethylamino)ethanol-1,1,2,2-D4
CAS 2733149-33-0
Molekylær formel C6 D4 H11 N O
Mængde 0.1 g
Synonym N,N-Diethylethanol-1,1,2,2-d4-amine, 2-(Diethylamino)ethanol-1,1,2,2-d4, 2-(Diethylamino)ethanol-1,1,2,2-D4
InChI formel InChI=1 S/C6H15NO/c1-3-7(4-2)5-6-8/h8H,3-6H2,1-2H3/i5D2,6D2
SMIL [2 H]C([2 H])(O)C([2 H])([2 H])N(CC)CC
IUPAC navn 1,1,2,2-tetradeuterio-2-(diethylamino)ethanol
Molekylvægt (g/mol) 121.214
Formel vægt 121.14 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity