Learn More
N,N-Diethylacetamide, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
N,N-Diethylacetamide was used to investigate the spectroscopic properties ofN,N-diethyl-2-[(4-substituted)phenylsulfinyl] acetamides. It is commonly used as drug solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N-Diethylacetamide was used to investigate the spectroscopic properties ofN,N-diethyl-2-[(4-substituted)phenylsulfinyl] acetamides. It is commonly used as drug solvent.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
N,N-Diethylacetamide was used to investigate the spectroscopic properties ofN,N-diethyl-2-[(4-substituted)phenylsulfinyl] acetamides. It is commonly used as drug solvent.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
Density | 0.92 |
Boiling Point | 182°C to 186°C |
Flash Point | 76°C (169°F) |
Refractive Index | 1.44 |
Quantity | 25 g |
Beilstein | 1209428 |
Solubility Information | Soluble in water. |
Formula Weight | 115.18 |
Percent Purity | 99% |
Chemical Name or Material | N,N-Diethylacetamide |