Få mere at vide
N,N’-Diacetyl-L-cystine Bis(tert-Butyl) Diester-D6, TRC
Artikelnummer. 30361519
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30361519 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361519 Leverandør Toronto Research Chemicals Leverandørnr. TRCD312517100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N'-Diacetyl-L-cystine Bis(tert-Butyl) Diester-D6
CAS 1356382-52-9
Molekylær formel C18 H26 D6 N2 O6 S2
Mængde 100 mg
Synonym N,N’-Diacetyl-L-cystine 1,1’-Bis(1,1-dimethylethyl) Ester-D6, Di-tert-butyl 3,3'-Disulfanediyl(2R,2'R)-bis(2-(acetamido-2,2,2-D3)propanoate), tert-Butyl (2R)-3-[(2R)-3-[(2-Methylpropan-2-yl)oxy]-3-oxo-2-[(2,2,2-trideuterioacetyl)amino]propyl]disulfanyl-2-[(2,2,2-trideuterioacetyl)amino]propanoate
InChI formel InChI=1S/C18H32N2O6S2/c1-11(21)19-13(15(23)25-17(3,4)5)9-27-28-10-14(20-12(2)22)16(24)26-18(6,7)8/h13-14H,9-10H2,1-8H3,(H,19,21)(H,20,22)/t13-,14-/m0/s1/i1D3,2D3
SMIL [2H]C([2H])([2H])C(=O)N[C@@H](CSSC[C@H](NC(=O)C([2H])([2H])[2H])C(=O)OC(C)(C)C)C(=O)OC(C)(C)C
Molekylvægt (g/mol) 442.62
Formel vægt 442.2078