Få mere at vide
N,N-Di(ethyl-2,2,2-d3)aniline, CDN
Artikelnummer. 30321367
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321367 0.05 g -
30318666 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321367 Leverandør CDN Leverandørnr. CDND55270.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Diethylaniline-D6 (N,N-di(ethyl-2,2,2-D3))
CAS 358731-12-1
Molekylær formel C10 D6 H9 N
Mængde 0.05 g
Synonym Benzenamine, N,N-di(ethyl-2,2,2-d3)- (9 CI), N,N-Di(ethyl-2,2,2-d3)aniline, N,N-Di(ethyl-2,2,2-d3)benzenamine, N,N-Diethylaniline-D6 (N,N-di(ethyl-2,2,2-D3))
InChI formel InChI=1 S/C10H15N/c1-3-11(4-2)10-8-6-5-7-9-10/h5-9 H,3-4H2,1-2H3/i1D3,2D3
SMIL [2 H]C([2 H])([2 H])CN(CC([2 H])([2 H])[2 H])c1ccccc1
IUPAC navn N,N-bis(2,2,2-trideuterioethyl)aniline
Molekylvægt (g/mol) 155.27
Formel vægt 155.158 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity