Få mere at vide
N-(n-Butanesulfonyl)-O-[4-(4-pyridinyl)-butyl]-(S)-tyrosine, TRC
Artikelnummer. 30004105
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004105 50 mg
30008056 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30004105 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69016050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 149490-61-9
Molekylær formel C22 H30 N2 O5 S
Molekylvægt (g/mol) 434.55
Synonym Tirofiban Impurity C
IUPAC navn (2S)-2-(butylsulfonylamino)-3-[4-(4-pyridin-4-ylbutoxy)phenyl]propanoic acid
SMIL CCCCS(=O)(=O)N[C@@H](Cc1ccc(OCCCCc2ccncc2)cc1)C(=O)O
Tekniske data
Kemisk navn eller materiale N-(n-Butanesulfonyl)-O-[4-(4-pyridinyl)-butyl]-(S)-tyrosine
InChI formel InChI=1S/C22H30N2O5S/c1-2-3-16-30(27,28)24-21(22(25)26)17-19-7-9-20(10-8-19)29-15-5-4-6-18-11-13-23-14-12-18/h7-14,21,24H,2-6,15-17H2,1H3,(H,25,26)/t21-/m0/s1
Mængde 50 mg
Formel vægt 434.1875