missing translation for 'onlineSavingsMsg'
Få mere at vide

N,N-Bis(2-hydroxyethyl)dodecanamide(>80%), TRC

Artikelnummer. 30482393
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482393 5 g
30469209 1 g
30475484 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30482393 Leverandør Toronto Research Chemicals Leverandørnr. TRCN0782355G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 120-40-1
Molekylær formel C16 H33 N O3
Molekylvægt (g/mol) 287.44
Synonym Dodecanamide, N,N-bis(2-hydroxyethyl)-, N,N-Bis(2-hydroxyethyl)dodecanamide, A 2000, AC 1000, AC 1000 (amide), Alkamide LE, Aminon L 02, Amisol LDE, Amisol LDE-G, Amisol SG, Amycol LDE, Antistatic B 2, Bis(2-hydroxyethyl)lauramide, Chemistat 2500, Chemstat LD 100, Clindrol 100L, Clindrol 200L, Clindrol Superamide 100L, Comperlan LD, Condensate PL, Crillon LDE, Dehydat 10, Denone 2863, Detergent 6501, Detergent 6502, Diethanolamine lauroylamide, Diethanollauramide, Duspar LA 2000, Emid 6511, Empilan LDE, Ethylan MLD, Hetamide ML, Incromide LR, Javachem A 2000, LA 2000, LDA, LDA (surfactant), LDE, Lalmin D, Lankrostat JP, Lauramide DEA, Lauramido DEA, Lauric acid diethanolamide, Lauric diethanolamide, Lauroyl diethanolamide, Lauroyldiethanolamine, Lauryl diethanolamide, Mackamide LL, Mackamide LLM, Mazamide LS 196, Micopol LDE, Monamid 150LW, Monamid 150LWA, N,N-Bis(2-hydroxyethyl)lauramide, N,N-Bis(2-hydroxyethyl)lauroylamide, N,N-Bis(2-hydroxyethyl)laurylamide, N,N-Bis(hydroxyethyl)lauramide, N,N-Bis(β-hydroxyethyl)lauramide, N,N-Di(2-hydroxyethyl)lauramide, N,N-Diethanoldodecanamide, N,N-Diethanollauramide, N,N-Diethanollauric acid amide, N,N-Diethylollauramide, N-Dodecanoyldiethanolamine, N-Lauroyldiethanolamine, Neo Duspar LA 2000P, Ninaer, Ninol 30LL, Ninol 50LL, Ninol 52LL, Ninol 55LL, Ninol 70SL, Ninol AA 62, Ninol AA 62 Extra, Nissan Stafoam DL, Onyxol 345, PUB 350, Pionin D 1110, Profan AA 62EX, Protamide L 80M, Rewomid DL 203S, Rewomid DLMS, Richamide 6310, Richamide STD, Schercomid SL-EX, Stafoam DL, Standamid LD, Standamid LDS, Standamid LDS-RV, Standarmi AC LDS-RV, Steinamid DL 203S, Stepan LDA, Stremid K, Super Amide L 9A, Super Amide L 9C, Surfactant 6501, Synotol L 60, Tohol N 230X, Trisophone PK, Unamide J 56, Varamide ML 1, Witcamide 5138, Witcamide 5195
IUPAC navn N,N-bis(2-hydroxyethyl)dodecanamide
SMIL CCCCCCCCCCCC(=O)N(CCO)CCO

Tekniske data

Kemisk navn eller materiale Lauryl Diethanolamide
Mængde 5 g
InChI formel InChI=1S/C16H33NO3/c1-2-3-4-5-6-7-8-9-10-11-16(20)17(12-14-18)13-15-19/h18-19H,2-15H2,1H3
Formel vægt 287.246
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.