Learn More
N,N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic Acid, Millipore™
Product Code. 10331671
Change view
Click to view available options
Quantity:
25 g
250 g
Unit Size:
250g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10331671 250 g 250g
10209400 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10331671 Supplier Millipore Supplier No. 1152280250
 Restricted Item
Forventet forsendelse: 26-11-2026
PKa (20°C)7.15, ≥99% (alkalimetric)
buffer substance BES
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10191-18-1
Specifications
Quantity 250 g
Solution Type Buffer
MDL Number MFCD00007533
Synonym N,N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid, BES, N,N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid
Molecular Weight (g/mol) 213.25
Grade Research
Chemical Name or Material N,N-Bis(2-Hydroxyethyl)-2-Aminoethanesulfonic Acid