Få mere at vide
N,N'-Bis[2-[[[5-[(dimethylamino)methyl]furan-2-yl]-methyl]sulphanyl]ethyl]-2-nitroethene-1,1-diamine, Mikromol™
Artikelnummer. 30050991
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050991 25 mg
17931956 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050991 Leverandør Mikromol Leverandørnr. MM0086.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N,N'-Bis[2-[[[5-[(dimethylamino)methyl]furan-2-yl]-methyl]sulphanyl]ethyl]-2-nitroethene-1,1-diamine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N'-Bis[2-[((5-[(dimethylamino)methyl]furan-2-yl)-methyl)sulphanyl]ethyl]-2-nitroethene-1,1-diamine
CAS 72126-78-4
Analyt- eller komponentnavne N,N'-Bis[2-[((5-[(dimethylamino)methyl]furan-2-yl)-methyl)sulphanyl]ethyl]-2-nitroethene-1,1-diamine
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C22 H35 N5 O4 S2
Certificeringer/compliance ISO 17025
Synonym Ranitidine Hydrochloride Imp. A (EP), Ranitidine Imp. A (EP), Ranitidine USP Related Compound B, Ranitidine USP RC B, N,N'-Bis[2-[[[5-[(dimethylamino)methyl]furan-2-yl]-methyl]sulphanyl]ethyl]-2-nitroethene-1,1-diamine, AH 20720X, Ph Eur Ranitidine Impurity A, BP Ranitidine Impurity A, USP Ranitidine Related Compound B, Ranitidine bis-compound (AH20720), Ranitidine Related Compound B, Ranitidine Hydrochloride Impurity A, Ranitidine Impurity A
InChI formel InChI=1S/C22H35N5O4S2/c1-25(2)13-18-5-7-20(30-18)16-32-11-9-23-22(15-27(28)29)24-10-12-33-17-21-8-6-19(31-21)14-26(3)4/h5-8,15,23-24H,9-14,16-17H2,1-4H3
SMIL CN(C)Cc1oc(CSCCNC(=C[N+](=O)[O-])NCCSCc2oc(CN(C)C)cc2)cc1
IUPAC navn 1-N,1-N'-bis[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-2-nitroethene-1,1-diamine
Molekylvægt (g/mol) 497.67
Mængde 25 mg
Formel vægt 497.213 g/mol