Learn More
Alfa Aesar™ N,N'-(1,2-Dihydroxyethylene)bisacrylamide, Electrophoresis Reagent, 97%
Brand:  Alfa Aesar™ J66899.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15400667
  • 700.89 DKK / 5g
Estimated Shipment date
from Supplier 17-01-2019
Add to basket
Description
Description
  • A cross-linker for polyacrylamide gels
  • Useful when recovering protein from polyacrylamide gels
Specifications
Chemical Identifiers
868-63-3
200.194
ZMLXKXHICXTSDM-UHFFFAOYSA-N
102501
C=CC(=O)NC(C(NC(=O)C=C)O)O
C8H12N2O4
MFCD00008624
n,n'-1,2-dihydroxyethylene bisacrylamide, dheba, 1,2-bisacrylamido-1,2-ethanediol, 2-propenamide, n,n'-1,2-dihydroxy-1,2-ethanediyl bis, n,n'-1,2-dihydroxyethane-1,2-diyl diacrylamide, n,n'-1,2-dihydroxy-1,2-ethanediyl bis acrylamide, n,n-1,2-dihydroxyethylene bisacrylamide, n-1,2-dihydroxy-2-prop-2-enoylamino ethyl prop-2-enamide, n-1,2-dihydroxy-2-prop-2-enamido ethyl prop-2-enamide, acmc-209qbw
N-[1,2-dihydroxy-2-(prop-2-enoylamino)ethyl]prop-2-enamide
Specifications
N,N'-(1,2-Dihydroxyethylene)bisacrylamide
141°C to 143°C
Electrophoresis Reagent
C8H12N2O4
1941875
ZMLXKXHICXTSDM-UHFFFAOYSA-N
N-[1,2-dihydroxy-2-(prop-2-enoylamino)ethyl]prop-2-enamide
102501
Powder
White to Cream
5g
868-63-3
MFCD00008624
n,n'-1,2-dihydroxyethylene bisacrylamide, dheba, 1,2-bisacrylamido-1,2-ethanediol, 2-propenamide, n,n'-1,2-dihydroxy-1,2-ethanediyl bis, n,n'-1,2-dihydroxyethane-1,2-diyl diacrylamide, n,n'-1,2-dihydroxy-1,2-ethanediyl bis acrylamide, n,n-1,2-dihydroxyethylene bisacrylamide, n-1,2-dihydroxy-2-prop-2-enoylamino ethyl prop-2-enamide, n-1,2-dihydroxy-2-prop-2-enamido ethyl prop-2-enamide, acmc-209qbw
C=CC(=O)NC(C(NC(=O)C=C)O)O
200.194
200.2
97%