Få mere at vide
N-Methylurea-d6, TRC
Artikelnummer. 30469754
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30469754 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469754 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33817710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methylurea-d6
CAS 56683-43-3
Molekylær formel C2 D6 N2 O
Mængde 10 mg
Synonym Urea-d3, methyl-d3- (9CI), N-Methylurea-d6, Methylurea-D6, Methyl-d3-urea-d3, 1-(Methyl-d3)urea-d3
InChI formel InChI=1S/C2H6N2O/c1-4-2(3)5/h1H3,(H3,3,4,5)/i1D3/hD3
SMIL [2H]N([2H])C(=O)N([2H])C([2H])([2H])[2H]
IUPAC navn 1,1,3-trideuterio-3-(trideuteriomethyl)urea
Molekylvægt (g/mol) 80.1188
Formel vægt 80.0857