Få mere at vide
N-Methyltopiramate, Mikromol™
Artikelnummer. 30049363
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30049363 25 mg
1 options
Artikelnummer. 30049363 Leverandør Mikromol Leverandørnr. MM1348.120025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Methyltopiramate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl Topiramate
CAS 97240-80-7
Analyt- eller komponentnavne N-Methyl Topiramate
Forsendelsestilstand Room Temperature
Molekylær formel C13H23NO8S
Certificeringer/compliance ISO 9001
Synonym 2,3:4,5-bis-O-(1-Methylethylidene)-beta-D-Fructopyranose 1-(N-Methylsulfamate), 2,3:4,5-bis-O-(1-methylethylidene)-beta-D-fructopyranose Methylsulfamate, 5H-Bis[1,3]dioxolo[4,5-b:4',5'-d]pyran beta-D-Fructopyranose deriv.
SMIL CNS(=O)(=O)OC[C@@]12OC[C@H]3OC(C)(C)O[C@H]3[C@@H]1OC(C)(C)O2
Molekylvægt (g/mol) 353.39