Få mere at vide
N-Methylthiophene-2-carboxamide, Mikromol™
Artikelnummer. 17982236
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17982236 100 mg
17941836 25 mg
30463583 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17982236 Leverandør Mikromol Leverandørnr. MM0247.04
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Methylthiophene-2-carboxamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methylthiophene-2-carboxamide
CAS 39880-77-8
Analyt- eller komponentnavne N-Methylthiophene-2-carboxamide
Forsendelsestilstand Room Temperature
Molekylær formel C6 H7 N O S
Certificeringer/compliance ISO 17025
Synonym N-Methylthiophene-2-carboxamide, Tenoxicam Imp. C (EP), Tenoxicam Impurity C
InChI formel InChI=1S/C6H7NOS/c1-7-6(8)5-3-2-4-9-5/h2-4H,1H3,(H,7,8)
SMIL CNC(=O)c1cccs1
IUPAC navn N-methylthiophene-2-carboxamide
Molekylvægt (g/mol) 141.19
Mængde 100 mg
Formel vægt 141.0248 g/mol