Få mere at vide
N-Methyltetrahydrofolic Acid, TRC
Artikelnummer. 30136324
Skift visning
Mængde:
10 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136324 1 g
30012852 100 mg
30008736 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136324 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3381451G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 134-35-0
Molekylær formel C20 H25 N7 O6
Molekylvægt (g/mol) 459.46
Synonym 5-Methyltetrahydrofolate
IUPAC navn (2S)-2-[[4-[(2-amino-5-methyl-4-oxo-1,6,7,8-tetrahydropteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
SMIL CN1C(CNc2ccc(cc2)C(=O)N[C@@H](CCC(=O)O)C(=O)O)CNC3=C1C(=O)N=C(N)N3
Tekniske data
Kemisk navn eller materiale 5-Methyltetrahydrofolic acid
Renhedsgrad noter HPLC
Mængde 1 g
InChI formel InChI=1S/C20H25N7O6/c1-27-12(9-23-16-15(27)18(31)26-20(21)25-16)8-22-11-4-2-10(3-5-11)17(30)24-13(19(32)33)6-7-14(28)29/h2-5,12-13,22H,6-9H2,1H3,(H,24,30)(H,28,29)(H,32,33)(H4,21,23,25,26,31)/t12?,13-/m0/s1
Formel vægt 459.1866
Procent renhed >95%