Få mere at vide
N-Methylsaccharin, Mikromol™
Artikelnummer. 17929544
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17929544 100 mg
17999904 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17929544 Leverandør Mikromol Leverandørnr. MM0084.13
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Methylsaccharin
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methylsaccharin
CAS 15448-99-4
Analyt- eller komponentnavne N-Methylsaccharin
Forsendelsestilstand Room Temperature
Molekylær formel C8 H7 N O3 S
Certificeringer/compliance ISO 17025
Synonym N-Methylsaccharin
InChI formel InChI=1S/C8H7NO3S/c1-9-8(10)6-4-2-3-5-7(6)13(9,11)12/h2-5H,1H3
SMIL CN1C(=O)c2ccccc2S1(=O)=O
IUPAC navn 2-methyl-1,1-dioxo-1,2-benzothiazol-3-one
Molekylvægt (g/mol) 197.21
Mængde 100 mg
Formel vægt 197.0147 g/mol