Få mere at vide
N-Methylperfluorooctanesulfonamidoethanol-d7 (~80%), TRC
Artikelnummer. 30482486
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482486 5 mg
30467433 1 mg
30476935 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482486 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3273475MG
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1265205-95-5
Molekylær formel C11HD7F17NO3S
Molekylvægt (g/mol) 564.26
Synonym N-(2-Hydroxyethyl)-N-methylperfluorooctanesulfonamide D7 (N-ethylene D4, N-methyl D3), N-(2-Hydroxyethyl)-N-methylperfluorooctanesulfonamide D7, N-Methylperfluorooctane Sulfonamidoethanol D7 (N-methyl D3, N-ethylene D4), 1-Octanesulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-(2-hydroxyethyl-1,1,2,2-d4)-N-(methyl-d3)- (ACI), 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluoro-N-(2-hydroxyethyl-1,1,2,2-d4)-N-(methyl-d3)-1-octanesulfonamide (ACI), D7-N-MeFOSE, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-(2-hydroxyethyl-1,1,2,2-d4)-N-(methyl-d3)-1-octanesulfonamide
IUPAC navn 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-(1,1,2,2-tetradeuterio-2-hydroxyethyl)-N-(trideuteriomethyl)octane-1-sulfonamide
SMIL [2H]C([2H])([2H])N(C([2H])([2H])C([2H])([2H])O)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Tekniske data
Kemisk navn eller materiale N-Methylperfluorooctanesulfonamidoethanol-d7
Mængde 5 mg
InChI formel InChI=1S/C11H8F17NO3S/c1-29(2-3-30)33(31,32)11(27,28)9(22,23)7(18,19)5(14,15)4(12,13)6(16,17)8(20,21)10(24,25)26/h30H,2-3H2,1H3/i1D3,2D2,3D2
Formel vægt 564.0393