Få mere at vide
N-Methylperfluorooctanesulfonamidoethanol (50μg/mL in Methanol), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-(2-Hydroxyethyl)-N-methylperfluorooctanesulfonamide |
| CAS | 24448-09-7 |
| Molekylær formel | C11 H8 F17 N O3 S |
| Mængde | 1 mL |
| Synonym | N-Methyl-N-(2-hydroxyethyl)perfluorooctane Sulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluoro-N-(2-hydroxyethyl)-N-methyl-1-octanesulfonamide, 2-(N-Methylperfluoro-1-octanesulfonamido)ethanol, MeFOSE Alcohol, N-(2-Hydroxyethyl)-N-methylperfluorooctanesulfonamide, N-MeFOSE, N-Methyl-N-ethanolperfluorooctanesulfonamide, N-Methylheptadecafluorooctanesulfonamidoethanol, N-Methylperfluorooctanesulfonamide ethyl alcohol, N-Methylperfluorooctanesulfonamidoethanol |
| InChI formel | InChI=1S/C11H8F17NO3S/c1-29(2-3-30)33(31,32)11(27,28)9(22,23)7(18,19)5(14,15)4(12,13)6(16,17)8(20,21)10(24,25)26/h30H,2-3H2,1H3 |
| SMIL | CN(CCO)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC navn | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-(2-hydroxyethyl)-N-methyloctane-1-sulfonamide |
| Molekylvægt (g/mol) | 557.22 |
| Formel vægt | 556.9953 |
| Vis mere |