Learn More
N-Methylindole-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Catalog Number p-4259019
Click to view available options
Quantity:
1 g
5 g
683229-61-0
C15H20BNO2
257.14
MCCCOWZSGBKRCG-UHFFFAOYSA-N
1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-indole, n-methylindole-3-boronic acid pinacol ester, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indole, n-methylindole-3-boronic acid, pinacol ester, 1-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 1-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborole-2-yl-1h-indole, 1h-indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
23058003
1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indole
B1(OC(C(O1)(C)C)(C)C)C2=CN(C3=CC=CC=C23)C
683229-61-0
C15H20BNO2
257.14
MCCCOWZSGBKRCG-UHFFFAOYSA-N
1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-indole, n-methylindole-3-boronic acid pinacol ester, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indole, n-methylindole-3-boronic acid, pinacol ester, 1-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 1-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborole-2-yl-1h-indole, 1h-indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
23058003
1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indole
B1(OC(C(O1)(C)C)(C)C)C2=CN(C3=CC=CC=C23)C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 106°C to 112°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 257.14
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material N-Methylindole-3-boronic acid pinacol ester
Show More Show Less
compliance-icons
Product Identifier
  • N-Methylindole-3-boronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.