missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
N-Methyliminodiacetic Acid, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Methyliminodiacetic Acid |
| CAS | 4408-64-4 |
| Molekylær formel | C5 H9 N O4 |
| Mængde | 100 mg |
| Synonym | Glycine, N-(carboxymethyl)-N-methyl- (9CI, ACI), Acetic acid, (methylimino)di- (6CI, 7CI, 8CI), N-(Carboxymethyl)-N-methylglycine (ACI), (Carboxymethyl-methyl-amino)-acetic acid, (Methylimino)diacetic acid, 2,2'-(Methylazanediyl)diacetic acid, 2,2'-Methyliminodiacetic acid, 2-[(Carboxymethyl)(methyl)amino]acetic acid, 2-[Carboxymethyl(methyl)amino]acetic acid, Methylimidodiacetic acid, MIDA, N-Methylaminodiacetic acid, N-Methyliminodiacetic acid, NSC 11773 |
| InChI formel | InChI=1S/C5H9NO4/c1-6(2-4(7)8)3-5(9)10/h2-3H2,1H3,(H,7,8)(H,9,10) |
| SMIL | CN(CC(=O)O)CC(=O)O |
| IUPAC navn | 2-[carboxymethyl(methyl)amino]acetic acid |
| Molekylvægt (g/mol) | 147.13 |
| Formel vægt | 147.0532 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?