Få mere at vide
N-Methyliminobis(methylenephosphonic acid), TRC
Artikelnummer. 30469344
Skift visning
Mængde:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469344 250 mg
30468848 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30469344 Leverandør Toronto Research Chemicals Leverandørnr. TRCM393780250MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyliminobis(methylenephosphonic acid)
CAS 5995-25-5
Molekylær formel C3H11NO6P2
Mængde 250 mg
Synonym ((methylazanediyl)bis(methylene))bis(phosphonic acid)
InChI formel InChI=1S/C3H11NO6P2/c1-4(2-11(5,6)7)3-12(8,9)10/h2-3H2,1H3,(H2,5,6,7)(H2,8,9,10)
SMIL O=P(O)(CN(CP(O)(O)=O)C)O
Molekylvægt (g/mol) 219.07