Få mere at vide
N-Methylfluoxetine Hydrochloride, Mikromol™
Artikelnummer. 17923546
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
17923546 25 mg
1 options
Artikelnummer. 17923546 Leverandør Mikromol Leverandørnr. MM0256.210025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Methylfluoxetine Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl Fluoxetine Hydrochloride
CAS 885101-12-2
Analyt- eller komponentnavne N-Methyl Fluoxetine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C18H21ClF3NO
Certificeringer/compliance ISO 9001
Synonym N,N-Dimethyl-γ-[4-(trifluoromethyl)phenoxy]benzenepropanamine Hydrochloride
SMIL CN(C)CCC(C1=CC=CC=C1)OC2=CC=C(C(F)(F)F)C=C2.Cl
Molekylvægt (g/mol) 359.81