Få mere at vide
N-Methyldiethanolamine-13C,d3 Hydrochloride, TRC
Artikelnummer. 30470776
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30470776 25 mg
30466865 5 mg
30479529 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470776 Leverandør Toronto Research Chemicals Leverandørnr. TRCM30162725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1781841-60-8
Molekylær formel C413CH10D3NO2.HCl
Molekylvægt (g/mol) 159.63
Synonym 2,2'-(Methylimino)bis[ethanol]-13C,d3 Hydrochloride, 2,2'-(Methylimino)diethanol-13C,d3 Hydrochloride, 2-[(2-Hydroxyethyl)(methyl)amino]ethanol-13C,d3 Hydrochloride, N,N-Bis(2-hydroxyethyl)methylamine-13C,d3 Hydrochloride, N,N-Di(2-hydroxyethyl)-N-methylamine-13C,d3 Hydrochloride, N,N-Di(2-hydroxyethyl)methanamine-13C,d3 Hydrochloride, N,N-Di(2-hydroxyethyl)methylamine-13C,d3 Hydrochloride, N-(2-Hydroxyethyl)-N-methylethanolamine-13C,d3 Hydrochloride, N-Methyl-N,N-bis(2-hydroxyethyl)amine-13C,d3 Hydrochloride, N-Methyl-N,N-diethanolamine-13C,d3 Hydrochloride, N-Methylbis(2-hydroxyethyl)amine-13C,d3 Hydrochloride, N-Methyliminodiethanol-13C,d3 Hydrochloride
IUPAC navn 2,2'-((methyl-13C-d3)azanediyl)bis(ethan-1-ol) hydrochloride
SMIL OCCN([13C]([2H])([2H])[2H])CCO.Cl
Tekniske data
Kemisk navn eller materiale N-Methyldiethanolamine-13C,D3 Hydrochloride
Mængde 25 mg
InChI formel InChI=1S/C5H13NO2.ClH/c1-6(2-4-7)3-5-8;/h7-8H,2-5H2,1H3;1H/i1+1D3;
Formel vægt 159.1