missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Methylcytisine, TRC

Artikelnummer. 17991275
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17991275 25 mg
17964195 10 mg
17930465 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17991275 Leverandør Toronto Research Chemicals Leverandørnr. TRCM29468025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 486-86-2
Molekylær formel C12 H16 N2 O
Molekylvægt (g/mol) 204.27
Synonym (-)-N-Methylcytisine, 1,5-Methano-8H-pyrido[1,2-a][1,5]diazocin-8-one, 1,2,3,4,5,6-hexahydro-3-methyl-, (1R,5S)-, 1,5-Methano-8H-pyrido[1,2-a][1,5]diazocin-8-one, 1,2,3,4,5,6-hexahydro-3-methyl-, (1R)-, Cytisine, 12-methyl- (8CI), Cytisine, N-methyl- (7CI), Cytisine, methyl- (6CI), (-)-N-Methylcytisine, (1R,5S)-1,2,3,4,5,6-Hexahydro-3-methyl-1,5-methano-8H-pyrido[1,2-a][1,5]diazocin-8-one, Caulophyllin, Caulophylline, N-Methylcytisine
SMIL CN1C[C@@H]2C[C@H](C1)C3=CC=CC(=O)N3C2

Specifications

Kemisk navn eller materiale N-Methylcytisine
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C12H16N2O/c1-13-6-9-5-10(8-13)11-3-2-4-12(15)14(11)7-9/h2-4,9-10H,5-8H2,1H3/t9-,10+/m0/s1
Formel vægt 204.1263
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.