missing translation for 'onlineSavingsMsg'
Learn More

N-Methylcaprolactam, 96%, Thermo Scientific Chemicals

Catalog Number p-7023831
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
2556-73-2
C7H13NO
127.187
MFCD00003263
ZWXPDGCFMMFNRW-UHFFFAOYSA-N
n-methylcaprolactam, 1-methylcaprolactam, n-methyl-.epsilon.-caprolactam, 2h-azepin-2-one, hexahydro-1-methyl, n-methyl-e-caprolactam, epsilon-caprolactam, n-methyl, 2h-azepin-2-one, hexahydro-n-methyl, hexahydro-n-methyl-2h-azepin-2-one, n-methyl-epsilon-caprolactam, n-methyl caprolactam
17369
1-methylazepan-2-one
CN1CCCCCC1=O
This item is not returnable. View return policy
2556-73-2
C7H13NO
127.187
MFCD00003263
ZWXPDGCFMMFNRW-UHFFFAOYSA-N
n-methylcaprolactam, 1-methylcaprolactam, n-methyl-.epsilon.-caprolactam, 2h-azepin-2-one, hexahydro-1-methyl, n-methyl-e-caprolactam, epsilon-caprolactam, n-methyl, 2h-azepin-2-one, hexahydro-n-methyl, hexahydro-n-methyl-2h-azepin-2-one, n-methyl-epsilon-caprolactam, n-methyl caprolactam
17369
1-methylazepan-2-one
CN1CCCCCC1=O

N-Methylcaprolactam was used to study the inhibition mechanism of poly-[N-vinylcaprolactam] (PVCAP) and its related compounds against hydrate formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylcaprolactam was used to study the inhibition mechanism of poly-[N-vinylcaprolactam] (PVCAP) and its related compounds against hydrate formation.

Solubility
Fully miscible with water.

Notes
Hygroscopic, store away from moisture. Store in a tightly closed container in a cool, dry, well-ventilated area. Store under dry inert gas.
TRUSTED_SUSTAINABILITY
Density 0.991
Boiling Point 106°C to 108°C (6 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.484
Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Fully miscible with water.
Formula Weight 127.18
Percent Purity 96%
Chemical Name or Material N-Methylcaprolactam

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'