Få mere at vide
N-Methylaniline Hydrochloride, TRC
Artikelnummer. 30470487
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30470487 1 g
30484031 10 g
30480519 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30470487 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2883081G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2739-12-0
Molekylær formel C7H9N . HCl
Molekylvægt (g/mol) 143.61
Synonym N-Methylanilinium Chloride, N-Methylbenzenamine Hydrochloride, Anilinomethane Hydrochloride
IUPAC navn N-methylaniline;hydrochloride
SMIL Cl.CNc1ccccc1
Tekniske data
Kemisk navn eller materiale N-Methylaniline Hydrochloride
Mængde 1 g
InChI formel InChI=1S/C7H9N.ClH/c1-8-7-5-3-2-4-6-7;/h2-6,8H,1H3;1H
Formel vægt 143.0502