Få mere at vide
N-Methylaniline-2,3,4,5,6-d5, TRC
Artikelnummer. 30481609
Skift visning
Mængde:
2.5 mg
5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30481609 5 mg -
30472787 2.5 mg 2.5mg
30481051 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30481609 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2882075MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 619319-96-9
Molekylær formel C7 D5 H4 N
Molekylvægt (g/mol) 112.1839
Synonym Benzen-d5-amine, N-methyl- (9CI)
IUPAC navn 2,3,4,5,6-pentadeuterio-N-methylaniline
SMIL [2H]c1c([2H])c([2H])c(NC)c([2H])c1[2H]
Tekniske data
Kemisk navn eller materiale N-Methylaniline-2,3,4,5,6-d5
Mængde 5 mg
InChI formel InChI=1S/C7H9N/c1-8-7-5-3-2-4-6-7/h2-6,8H,1H3/i2D,3D,4D,5D,6D
Formel vægt 112.1049