Få mere at vide
N-Methylaniline-2,3,4,5,6-d5, CDN
Artikelnummer. 30311900
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311900 0.1 g
30312949 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311900 Leverandør CDN Leverandørnr. CDND73680.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methylaniline-2,3,4,5,6-d5
CAS 619319-96-9
Molekylær formel C7 D5 H4 N
Mængde 0.1 g
Synonym Benzen-d5-amine, N-methyl- (9 CI)
InChI formel InChI=1 S/C7H9N/c1-8-7-5-3-2-4-6-7/h2-6,8 H,1H3/i2D,3 D,4 D,5 D,6 D
SMIL [2 H]c1c([2 H])c([2 H])c(NC)c([2 H])c1[2 H]
IUPAC navn 2,3,4,5,6-pentadeuterio-N-methylaniline
Molekylvægt (g/mol) 112.1839
Formel vægt 112.1049 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity