Få mere at vide
N-Methylacetamide-d7, CDN
Artikelnummer. 30309848
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30309848 5 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309848 Leverandør CDN Leverandørnr. CDND59535G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methylacetamide-D7
CAS 3669-74-7
Molekylær formel C3 D7 N O
Mængde 5 g
Synonym Acetamide-N,2,2,2-d4, N-(methyl-d3)- (7 CI, 8 CI, 9 CI), N-Methylacetamide-d7, N-Methylacetamide-D7, N-(Methyl-d3)acetamide-N,2,2,2-d4
InChI formel InChI=1 S/C3H7NO/c1-3(5)4-2/h1-2H3,(H,4,5)/i1D3,2D3/hD
SMIL [2 H]N(C(=O)C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn N,2,2,2-tetradeuterio-N-(trideuteriomethyl)acetamide
Molekylvægt (g/mol) 80.137
Formel vægt 80.097 g/mol
Procent renhed 99 atom % D, min 97% Chemical Purity