Få mere at vide
N-Methyl Pantoprazole-d3 (Mixture of 1 and 3 isomers), TRC
Artikelnummer. 30465628
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30465628 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30465628 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32586750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl Pantoprazole-d3 (Mixture of 1 and 3 isomers)
CAS 1190008-84-4
Molekylær formel C17H14D3F2N3O4S
Mængde 50 mg
Synonym 5-(Difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridinyl)methyl]sulfinyl]-1-(methyl-d3)-1H-benzimidazole
InChI formel InChI=1S/C17H17F2N3O4S/c1-22-13-5-4-10(26-16(18)19)8-11(13)21-17(22)27(23)9-12-15(25-3)14(24-2)6-7-20-12/h4-8,16H,9H2,1-3H3/i1D3
SMIL COC(C(OC)=CC=N1)=C1CS(C2=NC3=CC(OC(F)F)=CC=C3N2C([2H])([2H])[2H])=O
IUPAC navn 5-(difluoromethoxy)-2-(((3,4-dimethoxypyridin-2-yl)methyl)sulfinyl)-1-(methyl-d3)-1H-benzo[d]imidazole
Molekylvægt (g/mol) 400.41
Formel vægt 400.11