Få mere at vide
N-Methyl Palbociclib Hydrochloride, TRC
Artikelnummer. 30481485
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481485 100 mg
30465005 50 mg
30481191 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30481485 Leverandør Toronto Research Chemicals Leverandørnr. TRCM304233100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 850918-91-1
Molekylær formel C25H31N7O2 . xHCl
Molekylvægt (g/mol) 498
Synonym 6-Acetyl-8-cyclopentyl-5-methyl-2-[[5-(4-methyl-1-piperazinyl)-2-pyridinyl]amino]-pyrido[2,3-d]pyrimidin-7(8H)-one Hydrochloride
SMIL Cl.CN1CCN(CC1)c2ccc(Nc3ncc4C(=C(C(=O)C)C(=O)N(C5CCCC5)c4n3)C)nc2
Tekniske data
Kemisk navn eller materiale N-Methyl Palbociclib Hydrochloride
Mængde 100 mg
InChI formel InChI=1S/C25H31N7O2.ClH/c1-16-20-15-27-25(28-21-9-8-19(14-26-21)31-12-10-30(3)11-13-31)29-23(20)32(18-6-4-5-7-18)24(34)22(16)17(2)33;/h8-9,14-15,18H,4-7,10-13H2,1-3H3,(H,26,27,28,29);1H