Få mere at vide
N-Methyl-N-(trimethylsilyl)trifluoroacetamide-d9, TRC
Artikelnummer. 30483868
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30483868 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483868 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33112710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl-N-(trimethylsilyl)trifluoroacetamide-d9 (MSTFA-d9)
CAS 945623-67-6
Molekylær formel C6 2H9 H3 F3 N O Si
Mængde 10 mg
InChI formel InChI=1S/C6H12F3NOSi/c1-10(12(2,3)4)5(11)6(7,8)9/h1-4H3/i2D3,3D3,4D3
SMIL [2H]C([2H])([2H])[Si](N(C)C(=O)C(F)(F)F)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 2,2,2-trifluoro-N-methyl-N-[tris(trideuteriomethyl)silyl]acetamide
Molekylvægt (g/mol) 208.3
Formel vægt 208.1205