Få mere at vide
N-Methyl-N'-(pyridin-2-yl)ethanediamide, Mikromol™
Artikelnummer. 17981826
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17981826 100 mg
17974286 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17981826 Leverandør Mikromol Leverandørnr. MM0247.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Methyl-N'-(pyridin-2-yl)ethanediamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl-N'-(pyridin-2-yl)ethanediamide
CAS 52781-01-8
Analyt- eller komponentnavne N-Methyl-N'-(pyridin-2-yl)ethanediamide
Forsendelsestilstand Room Temperature
Molekylær formel C8 H9 N3 O2
Certificeringer/compliance ISO 17025
Synonym N1-Methyl-N2-2-pyridinylethanediamide, Ethanediamide, N-methyl-N'-2-pyridinyl- (9CI), N1-Methyl-N2-2-pyridinylethanediamide, N-Methyl-N'-(pyridin-2-yl)-ethanediamide
InChI formel InChI=1S/C8H9N3O2/c1-9-7(12)8(13)11-6-4-2-3-5-10-6/h2-5H,1H3,(H,9,12)(H,10,11,13)
SMIL CNC(=O)C(=O)Nc1ccccn1
IUPAC navn N-methyl-N'-pyridin-2-yloxamide
Molekylvægt (g/mol) 179.18
Mængde 100 mg
Formel vægt 179.179 g/mol