Få mere at vide
N-Methyl-N-nitroso Urea (Wetted with water >20%), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Methyl-N-nitrosourea |
| CAS | 684-93-5 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C2 H5 N3 O2 |
| Mængde | 5 g |
| Synonym | Urea, N-methyl-N-nitroso- (9CI, ACI), N-Methyl-N-nitrosourea (ACI), Urea, 1-methyl-1-nitroso- (6CI, 8CI), 1-Methyl-1-nitrosourea, 1-Nitroso-1-methylurea, Methylnitrosourea, MNU, N-Nitroso-N-methylcarbamide, N-Nitroso-N-methylurea, Nitrosomethylurea, NMH, NMM, NMU, NSC 23909 |
| InChI formel | InChI=1S/C2H5N3O2/c1-5(4-7)2(3)6/h1H3,(H2,3,6) |
| SMIL | CN(N=O)C(=O)N |
| IUPAC navn | 1-methyl-1-nitrosourea |
| Molekylvægt (g/mol) | 103.08 |
| Vis mere |