Få mere at vide
N-Methyl-N,N-bis(2-pyridylethyl)amine-d3, TRC
Artikelnummer. 30473541
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473541 50 mg
30466692 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473541 Leverandør Toronto Research Chemicals Leverandørnr. TRCM29421750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl-N,N-bis(2-pyridylethyl)amine-d3
CAS 244094-71-1
Molekylær formel C15H16D3N3
Mængde 50 mg
Synonym N-(Methyl-d3)-N-[2-(2-pyridinyl)ethyl]-2-pyridineethanamine, (Methyl-d3)bis(2-pyridylethyl)amine, N-(Methyl-d3)bis[2-(2-pyridylethyl)]amine, NSC 19005-d3
InChI formel InChI=1S/C15H19N3/c1-18(12-8-14-6-2-4-10-16-14)13-9-15-7-3-5-11-17-15/h2-7,10-11H,8-9,12-13H2,1H3/i1D3
SMIL [2H]C([2H])([2H])N(CCc1ccccn1)CCc2ccccn2
IUPAC navn 2-pyridin-2-yl-N-(2-pyridin-2-ylethyl)-N-(trideuteriomethyl)ethanamine
Molekylvægt (g/mol) 244.35
Formel vægt 244.1767