Få mere at vide
N-Methyl Moxifloxacin, TRC
Artikelnummer. 30133581
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133581 10 mg
30003796 50 mg
30005095 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133581 Leverandør Toronto Research Chemicals Leverandørnr. TRCM22182410MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 721970-37-2
Molekylær formel C22 H26 F N3 O4
Molekylvægt (g/mol) 415.46
Synonym 3-Quinolinecarboxylic acid, 1-cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aS,7aS)-octahydro-1-methyl-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-, 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aS,7aS)-octahydro-1-methyl-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic acid, N-Methylmoxifloxacin, Moxifloxacin EP Impurity F (N-Methyl Moxifloxacin)
IUPAC navn 7-[(4aS,7aS)-1-methyl-3,4,4a,5,7,7a-hexahydro-2H-pyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid
SMIL COc1c(N2C[C@@H]3CCCN(C)[C@@H]3C2)c(F)cc4C(=O)C(=CN(C5CC5)c14)C(=O)O
Tekniske data
Kemisk navn eller materiale N-Methyl Moxifloxacin
Mængde 10 mg
InChI formel InChI=1S/C22H26FN3O4/c1-24-7-3-4-12-9-25(11-17(12)24)19-16(23)8-14-18(21(19)30-2)26(13-5-6-13)10-15(20(14)27)22(28)29/h8,10,12-13,17H,3-7,9,11H2,1-2H3,(H,28,29)/t12-,17+/m0/s1
Formel vægt 415.1907